Gallium triiodide, ultra dry ≥99.999% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Ampuled under argon
47148.03
47148.06
47148.14
47148.03EA
3030
CZK
47148.03
47148.06
47148.14
Gallium triiodide, ultra dry ≥99.999% (metals basis)
Gallium triiodide
Purity: 99.999%
CAS No.: 13450-91-4
Molecular Formula: GaI3
Formula Weight: 450.43
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.15
Sensitivity: Moisture and Light Sensitive
Appearance: Yellow-green
Melting Point: 212°
Boiling Point: 345° subl.
UN No.: UN3260
MDL No.: MFCD00016107
Vzorec:
GaI₃ MW: 450,44 g/mol Bod varu: 345 °C (subl.) Bod tání: 212 °C Hustota: 4,15 Storage Temperature: Ambient |
MDL Number:
MFCD00016107 CAS číslo: 13450-91-4 EINECS: 236-611-6 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...