Toluen, anhydrous ≥99.8%, over molecular sieves
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Packaged under argon in resealable ChemSeal™ bottles.
47136.AE
47136.K2
47136.AEEA
1430
CZK
47136.AE
47136.K2
Toluen, anhydrous ≥99.8%, over molecular sieves
Toluen
Purity: 99.8%
CAS No.: 108-88-3
Molecular Formula: C7H8
Formula Weight: 92.14
Flash Point: 4°(40°F)
Physical Form: Liquid
Density: 0.867
Refractive Index: 1.4967
Appearance: Colourless
Odour: Petroleum-like
Melting Point: -93°
Boiling Point: 111°
Beilstein Reference: 635760
UN No.: UN1294
MDL No.: MFCD00008512
Vzorec:
C₆H₅CH₃ Bod varu: 111 °C Bod tání: –93 °C Hustota: 0,867 Bod vzplanutí: 4 °C (40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS číslo: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 14,9529 |