Tisk…

Nickelbis(trifluoromethylsulphonyl)imide

Dodavatel: Thermo Scientific
Nebezpečí

47102.MD 47102.03 47102.06
47102.MDEA 1710 CZK
47102.MD 47102.03 47102.06
Nickelbis(trifluoromethylsulphonyl)imide
Nickelbis(trifluoromethylsulphonyl)imide

CAS No.: 207861-63-0 Molecular Formula: C4F12N2NiO8S4 Formula Weight: 618.96 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Green UN No.: UN3261 MDL No.: MFCD23380174

Vzorec: C₄F₁₂N₂NiO₈S₄
MW: 618,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380174
CAS číslo: 207861-63-0
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR