Polyethylene glycol dimethacrylate, M.W. 8,000
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
PEGDMA, Polyethylene Glycol Dimethacrylate average Mn 550, PEG dimethacrylate, Polyethylene Glycol Dimethacrylate
CAS No.: 25852-47-5
Molecular Formula: CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Storage Temperature: -30°C to -10°C
Physical Form: Granular solid
Sensitivity: Moisture and Light Sensitive
Appearance: White to off-white
MDL No.: MFCD00081877
Vzorec:
(C₂H₄O)nC₈H₁₀O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00014932 CAS číslo: 25852-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...