Polyoxyethylene bis(amine), M.W. 1,000
Dodavatel: Thermo Fisher Scientific
46833.MD
46833.03
46833.06
46833.MDEA
1740
CZK
46833.MD
46833.03
46833.06
Polyoxyethylene bis(amine), M.W. 1,000
Polyoxyethylene bis(amine)
CAS No.: 24991-53-5
Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2
Physical Form: Granular solid
Sensitivity: Air Sensitive
Appearance: White to off-white
MDL No.: MFCD00132004
Vzorec:
H₂NCH₂CH₂(OCH₂CH₂)nNH₂ Storage Temperature: Ambient |
MDL Number:
MFCD00132004 CAS číslo: 24991-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...