Tisk…
Paladium 1% on carbon, granules, reduced
Dodavatel: Thermo Fisher Scientific
46462.14 46462.22 46462.36
46462.14EA 4530 CZK
46462.14 46462.22 46462.36
Paladium 1% on carbon, granules, reduced
Palladium
CAS No.: 7440-05-3 Physical Form: 3mm (0.13in) Pellets MDL No.: MFCD00011167
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS číslo: 7440-05-3