Tisk…

Polyester acrylate/methacrylate PEAM-1769

Dodavatel: Thermo Scientific
Varování

46351.14 46351.22
46351.14EA 3540 CZK
46351.14 46351.22
Polyester acrylate/methacrylate PEAM-1769
Polyester acrylate/methacrylate

CAS No.: 921214-61-1 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Amber


Applications: Recommended for use as a base resin or an additive to reduce stress

MW: 645 g/mol
Storage Temperature: Refrigerator
CAS číslo: 921213-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR