Ammonium pentaborate tetrahydrate 98% 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ammonium borate tetrahydrate
46323.A3
46323.36
46323.A3EA
8690
CZK
46323.A3
46323.36
Ammonium pentaborate tetrahydrate 98% 98%
Ammonium pentaborate tetrahydrate
Purity: 99+%
CAS No.: 12046-04-7
Molecular Formula: H8B10N2O16.4H2O
Formula Weight: 472.23 (400.17anhy)
Physical Form: Powder
Density: 1.58
Appearance: White
Melting Point: 110°
MDL No.: MFCD13176621
Vzorec:
H₄B₅NO₈·4H₂O MW: 272.15 g/mol Bod tání: 110 °C Hustota: 1.58 Storage Temperature: Ambient |
MDL Number:
MFCD13176621 CAS číslo: 12046-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...