Oxid hlinitý superaktivovaný kyselý Grade I
Dodavatel: Thermo Fisher Scientific
46065.A1
46065.30
46065.A1EA
8930
CZK
46065.A1
46065.30
Oxid hlinitý superaktivovaný kyselý Grade I
Aluminium oxide superactivated acidic
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 50-200 Micron APS Powder
Refractive Index: 1.765
Appearance: White
Odour: Odourless
Melting Point: 2045°
Boiling Point: 2980°
MDL No.: MFCD00003424
Vzorec:
Al₂O₃ MW: 101.96 g/mol Bod varu: 2980 °C Bod tání: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS číslo: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...