Tisk…

Oxid hlinitý kyselý HPLC flash grade

Dodavatel: Thermo Fisher Scientific

45956.22 45956.36
45956.22EA 1440 CZK
45956.22 45956.36
Oxid hlinitý kyselý HPLC flash grade
Oxid hlinitý kyselý

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 32-63 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Vzorec: Al₂O₃
Bod varu: 2980 °C
Bod tání: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR