Zeolite mordenite, sodium
Dodavatel: Thermo Fisher Scientific
45876.14
45876.22
45876.36
45876.14EA
1540
CZK
45876.14
45876.22
45876.36
Zeolite mordenite, sodium
Zeolite mordenite, sodium
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 425 m2/g, 13:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics
Vzorec:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS číslo: 1318-02-1 EINECS: 215-283-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...