Tisk…

Zeolite β, ammonium

Dodavatel: Thermo Scientific

45874.14 45874.22 45874.36
45874.14EA 1390 CZK
45874.14 45874.22 45874.36
Zeolite β, ammonium
Zeolite beta, ammonium

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 710 m2/g, 38:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)

Vzorec: SiO₂:Al₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS číslo: 1318-02-1
EINECS: 215-283-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR