Zeolite Y, ammonium
Dodavatel: Thermo Scientific
45869.14
45869.22
45869.36
45869.14EA
1410
CZK
45869.14
45869.22
45869.36
Zeolite Y, ammonium
Zeolite Y, ammonium
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 730 m2/g, 12:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products
Vzorec:
SiO₂:Al₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00132601 CAS číslo: 1318-02-1 EINECS: 215-283-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...