Bis(tri-o-tolylphosphine)palladium(0) (14,9% Pd)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
45814.MD
45814.03
45814.MDEA
3140
CZK
45814.MD
45814.03
Bis(tri-o-tolylphosphine)palladium(0) (14,9% Pd)
Bis(tri-o-tolylphosphine)palladium(0)
CAS No.: 69861-71-8
Molecular Formula: C42H42P2Pd
Formula Weight: 715.16
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Off-white to yellow
Melting Point: 220-225°
MDL No.: MFCD12911908
Vzorec:
C₄₂H₄₂P₂Pd Bod tání: 220…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD12911908 CAS číslo: 69861-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...