Bis(tricyclohexylphosphine)palladium(0) ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
45792.MD
45792.03
45792.MDEA
4330
CZK
45792.MD
45792.03
Bis(tricyclohexylphosphine)palladium(0) ≥97%
Bis(tricyclohexylphosphine)palladium(0)
Purity: 97% min
CAS No.: 33309-88-5
Molecular Formula: C36H66P2Pd
Formula Weight: 667.29
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white
Odour: Odourless
MDL No.: MFCD01073796
Vzorec:
C₃₆H₆₆P₂Pd MW: 667.292 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01073796 CAS číslo: 33309-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...