Paladium 5% on activated carbon wetted with ca. 50% water, powder Type A405023-5
Dodavatel: Thermo Fisher Scientific
45051.06
45051.22
45051.14
45051.06EA
2840
CZK
45051.06
45051.22
45051.14
Paladium 5% on activated carbon wetted with ca. 50% water, powder Type A405023-5
Palladium
CAS No.: 7440-05-3
Physical Form: Notes: Sold on a dry weight basis
Odour: Odourless
MDL No.: MFCD00011167
Vzorec:
Pd Storage Temperature: Ambient |
MDL Number:
MFCD03457879 CAS číslo: 7440-05-3 |