Tetraamminepalladium(II) sulphate (≥37,4% Pd) ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
44956.14
44956.06
44956.03
44956.14EA
96260
CZK
44956.14
44956.06
44956.03
Tetraamminepalladium(II) sulphate (≥37,4% Pd) ≥99.9% (metals basis)
Tetraamminepalladium(II) sulphate
Purity: 99.9%
CAS No.: 13601-06-4
Molecular Formula: (NH3)4PdSO4
Formula Weight: 270.61
Physical Form: Crystalline
MDL No.: MFCD09953444
Vzorec:
(NH₃)₄PdSO₄ MW: 270,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953444 CAS číslo: 13601-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...