Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Dodavatel: Thermo Scientific
|
Varování
|
44955.06
44955.14
44955.03
44955.06EA
18190
CZK
44955.06
44955.14
44955.03
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
CAS No.: 16971-33-8
Molecular Formula: C55H46ClOP3Ru
Formula Weight: 952.41
Physical Form: Crystalline
Sensitivity: Moisture and Light Sensitive
Appearance: Gray
Melting Point: 204-206° dec.
MDL No.: MFCD00049804
Vzorec:
C₅₅H₄₅ClOP₃Ru MW: 951,405 g/mol Bod tání: 204…206 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00049804 CAS číslo: 16971-33-8 EINECS: 241-051-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...