Antimony tin oxide ≥99.5%, NanoArc®
Dodavatel: Thermo Fisher Scientific
Varování
|
44930.14
44930.22
44930.36
44930.14EA
1330
CZK
44930.14
44930.22
44930.36
Antimony tin oxide ≥99.5%, NanoArc®
Antimony tin oxide
Purity: 99.5%
Molecular Formula: Sb2O5:SnO2 15:85 wt%
Physical Form: 13-22nm APS Powder, S.A. 42-72m2/g
UN No.: UN1549
MDL No.: MFCD00799153
Applications: Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge
Vzorec:
Sb₂O₅:SnO₂₁₅:₈₅% Storage Temperature: Ambient |
MDL Number:
MFCD00799153 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...