3-Trimethylsilylpropynal ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-(Trimethylsilyl)-2-propynal
Packaged under argon.
44763.03
44763.06
44763.03EA
2880
CZK
44763.03
44763.06
3-Trimethylsilylpropynal ≥97%
3-Trimethylsilylpropynal
Purity: 97%
CAS No.: 2975-46-4
Molecular Formula: C6H10OSi
Formula Weight: 126.23
Storage Temperature: 2°C to 8°C
Flash Point: 29°(84°F)
Physical Form: Liquid, Packaged under argon
Density: 0.86
Refractive Index: 1.4435
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless to yellow
Boiling Point: 58-60°/37.5mm
UN No.: UN1993
MDL No.: MFCD01103709
Vzorec:
C₆H₁₀OSi Bod varu: 58…60 °C (37,5 mmHg) Hustota: 0,86 Bod vzplanutí: 29 °C (84 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD01103709 CAS číslo: 2975-46-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...