Ruthenium 2% on alumina, pellets
Dodavatel: Thermo Scientific
44575.14
44575.22
44575.36
44575.14EA
4190
CZK
44575.14
44575.22
44575.36
Ruthenium 2% on alumina, pellets
Ruthenium
CAS No.: 7440-18-8
Physical Form: S.A. nominally 200m2/g
Odour: Odourless
MDL No.: MFCD00011207
Vzorec:
Ru Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS číslo: 7440-18-8 EINECS: 231-127-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...