Polyphenylmethylsiloxane, M.W. 2500-2700
Dodavatel: Thermo Scientific
44569.18
44569.30
44569.A1
44569.18EA
1590
CZK
44569.18
44569.30
44569.A1
Polyphenylmethylsiloxane, M.W. 2500-2700
Polyphenylmethylsiloxane
CAS No.: 9005-12-3
Molecular Formula: (C6H5(CH3)SiO2)2
Flash Point: 300°(572°F)
Physical Form: Liquid
MDL No.: MFCD00146731
Vzorec:
(C₆H₅(CH₃)SiO₂)₂ Bod vzplanutí: 300 °C (572 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00146731 CAS číslo: 9005-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...