Potassium hexachlororuthenate(III) (≥23% Ru) ≥99.99% (metals basis), Premion®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
44547.03
44547.06
44547.03EA
3470
CZK
44547.03
44547.06
Potassium hexachlororuthenate(III) (≥23% Ru) ≥99.99% (metals basis), Premion®
Potassium hexachlororuthenate(III)
Purity: 99.99%
CAS No.: 25443-63-4
Molecular Formula: K3RuCl6
Formula Weight: 431.09
Physical Form: Crystalline
MDL No.: MFCD00011390
Vzorec:
K₃RuCl₆ Storage Temperature: Ambient |
MDL Number:
MFCD00011390 CAS číslo: 25443-63-4 EINECS: 246-983-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...