Tisk…
Platina 10% on alumina, reduced
Dodavatel: Thermo Fisher Scientific
Synonyms: Platinium
44494.14 44494.22
44494.14EA 18640 CZK
44494.14 44494.22
Platina 10% on alumina, reduced
Platina
CAS No.: 7440-06-4 Physical Form: Granular MDL No.: MFCD00011179
Vzorec: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4