Tisk…

(S)-(+)-Methyl-3-hydroxy-2-methylpropionate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (S)-Methyl 3-hydroxy-2-methylpropanoate

44483.04 44483.09
44483.04EA 3640 CZK
44483.04 44483.09
(S)-(+)-Methyl-3-hydroxy-2-methylpropionate ≥98%
(S)-(+)-Methyl-3-hydroxy-2-methylpropionate

Purity: 99% CAS No.: 80657-57-4 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 81°(178°F) Physical Form: Liquid Density: 1.07 Refractive Index: 1.4250 Appearance: Colourless Boiling Point: 74°/10mm Beilstein Reference: 1720830 Optical Rotation: +27° (c=4 in methanol) MDL No.: MFCD00064520

Vzorec: C₅H₁₀O₃
MW: 118,132 g/mol
Bod varu: 74 °C (10 mmHg)
Hustota: 1,07
Bod vzplanutí: 81 °C (178 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064520
CAS číslo: 80657-57-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR