Tisk…
Platina 5% graphite Type 286
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Platinium
44365.06 44365.14 44365.22
44365.06EA 3510 CZK
44365.06 44365.14 44365.22
Platina 5% graphite Type 286
Platina
CAS No.: 7440-06-4 Physical Form: Powder Appearance: Dark grey Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179
Applications: Material for gas sensors
Vzorec: Pt
MW: 195.08
Bod varu: 3827
Bod tání: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1 III
Merck Index: 12,07689