Tisk…
Paladium 10% on activated carbon powder standard wetted with ca. 50% water, reduced Type 58
Dodavatel: Thermo Fisher Scientific
44350.06 44350.14 44350.22
44350.06EA 5670 CZK
44350.06 44350.14 44350.22
Paladium 10% on activated carbon powder standard wetted with ca. 50% water, reduced Type 58
Palladium
Appearance: Black Odour: Odourless
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic nitrites to 1° amines
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS číslo: 7440-05-3