Tisk…
Platina 5% on alumina, powder, reduced
Dodavatel: Thermo Fisher Scientific
Synonyms: Platinium
44222.14 44222.22 44222.06
44222.14EA 16410 CZK
44222.14 44222.22 44222.06
Platina 5% on alumina, powder, reduced
Platina
CAS No.: 7440-06-4 MDL No.: MFCD03458043
Vzorec: Pt
MW: 195.08
Bod varu: 3827
Bod tání: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
Merck Index: 12,07689