Tisk…
Paladium 5% on activated carbon powder standard wetted with ca. 50% water acidic catalyst, reduced
Dodavatel: Thermo Fisher Scientific
44142.09 44142.18 44142.30
44142.09EA 5670 CZK
44142.09 44142.18 44142.30
Paladium 5% on activated carbon powder standard wetted with ca. 50% water acidic catalyst, reduced
Palladium
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
Physical Form: 900-1100 m²/g Odor: Odourless
Applications: Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3