Tisk…

Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Dodavatel: Thermo Fisher Scientific

44123.02 44123.04
44123.02EA 8140 CZK
44123.02 44123.04
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

CAS No.: 36309-88-3 Molecular Formula: C72H48Cl2N6Ru Formula Weight: 1169.20 Physical Form: Powder MDL No.: MFCD03095387


Applications: Redox indicator, oxygen sensor

Vzorec: C₇₂H₄₈Cl₂N₆Ru
MW: 1169.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095387
CAS číslo: 36309-88-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR