Cobalt(II) sulphamate hydrate
Dodavatel: Thermo Scientific
|
Varování
|
43906.22
43906.36
43906.A3
43906.22EA
5360
CZK
43906.22
43906.36
43906.A3
Cobalt(II) sulphamate hydrate
Cobalt(II) sulphamate hydrate
CAS No.: 14017-41-5
Molecular Formula: Co(SO3NH2)2.xH2O
Formula Weight: 251.11(anhy)
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00049877
Vzorec:
CoH₄N₂O₆S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00049877 CAS číslo: 14017-41-5 EINECS: 237-834-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...