Zinc 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
43903.AC
43903.AE
43903.ACEA
3920
CZK
43903.AC
43903.AE
Zinc 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol
Zinc 2-methoxyethoxide
CAS No.: 129918-15-6
Molecular Formula: C6H14O2Zn
Formula Weight: 215.56
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
Boiling Point: 124°
UN No.: UN1188
MDL No.: MFCD00156529
Vzorec:
C₆H₁₄O₂Zn Bod varu: 124 °C Hustota: 0,96 Bod vzplanutí: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00156529 CAS číslo: 129918-15-6 UN: 1188 ADR: 3,III |