Magnesium tetraborate ≥99%
Dodavatel: Thermo Fisher Scientific
43892.14
43892.22
43892.14EA
4710
CZK
43892.14
43892.22
Magnesium tetraborate ≥99%
Magnesium tetraborate
Purity: 99%
CAS No.: 12007-62-4
Molecular Formula: MgB4O7
Formula Weight: 179.54
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD02688001
Vzorec:
MgB₄O₇ Storage Temperature: Ambient |
MDL Number:
MFCD02688001 CAS číslo: 12007-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...