Sulphobromophthalein sodium salt hydrate
Dodavatel: Thermo Fisher Scientific
43878.03
43878.06
43878.14
43878.03EA
996
CZK
43878.03
43878.06
43878.14
Sulphobromophthalein sodium salt hydrate
Sulphobromophthalein sodium salt hydrate
CAS No.: 71-67-0
Molecular Formula: C20H8Br4O10S2Na2.xH2O
Formula Weight: 837.98(anhy)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
MDL No.: MFCD00150017
Vzorec:
C₂₀H₈Br₄O₁₀S₂Na₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150017 CAS číslo: 71-67-0 EINECS: 200-761-0 Merck Index: 14,8954 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...