catalyst support
Dodavatel: Thermo Fisher Scientific
43815.22
43815.A4
43815.36
43815.22EA
2980
CZK
43815.22
43815.A4
43815.36
catalyst support
Zirconium oxide
Physical Form: 1/8" Pellets
Odour: Odourless
MDL No.: MFCD00011310
Applications: Catalyst carrier
Vzorec:
OZr MW: 107.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011310 CAS číslo: 12036-01-0 EINECS: 215-227-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...