Leuco malachite green
Dodavatel: Thermo Fisher Scientific
Synonyms:
4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-(Phenylmethylene)bis(N,N-dimethylaniline), 4,4'-Tetramethyldiaminotriphenylmethane, 4,4'-Benzylidenebis(N,N-dimethylaniline), Leucomalachite green
43758.09
43758.04
43758.09EA
3750
CZK
43758.09
43758.04
Leuco malachite green
Leuco malachite green
CAS No.: 129-73-7
Molecular Formula: C23H26N2
Formula Weight: 330.47
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 100-102°
Beilstein Reference: 2140506
MDL No.: MFCD00008315
Vzorec:
C₂₃H₂₆N₂ MW: 330.48 g/mol Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008315 CAS číslo: 129-73-7 EINECS: 204-961-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...