4-Bromomethyl-6,7-dimethoxycoumarin ≥95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
43402.04
43402.77
43402.04EA
12940
CZK
43402.04
43402.77
4-Bromomethyl-6,7-dimethoxycoumarin ≥95%
4-Bromomethyl-6,7-dimethoxycoumarin
Purity: 95%
CAS No.: 88404-25-5
Molecular Formula: C12H11BrO4
Formula Weight: 299.12
Physical Form: Powder
Melting Point: 200-203°
Beilstein Reference: 4750487
UN No.: UN3261
MDL No.: MFCD00011570
Applications: Fluorescent labeling reagent. Used in HPLC for the detection of carboxylic acids
Vzorec:
C₁₂H₁₁BrO₄ MW: 299,12 g/mol Bod tání: 200…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011570 CAS číslo: 88404-25-5 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...