Tisk…

Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 4-aminothiophene-3-carboxylate hydrochloride

43384.03 43384.06
43384.03EA 2860 CZK
43384.03 43384.06
Methyl-4-aminothiophene-3-carboxylate hydrochloride ≥97%
Methyl-4-aminothiophene-3-carboxylate hydrochloride

Purity: 97+% CAS No.: 39978-14-8 Molecular Formula: C6H7NO2S.HCl Formula Weight: 193.65 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Brown Melting Point: 199-200° dec. MDL No.: MFCD00068149

Vzorec: C₆H₈ClNO₂S
MW: 193,65 g/mol
Bod tání: 199…200 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00068149
CAS číslo: 39978-14-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR