Tris(dimethylamino)aluminum dimer
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Packaged under argon.
43237.03
43237.06
43237.03EA
5430
CZK
43237.03
43237.06
Tris(dimethylamino)aluminum dimer
Tris(dimethylamino)aluminum dimer
CAS No.: 32093-39-3
Molecular Formula: C12H36Al2N6
Formula Weight: 318.42
Flash Point: 21°(70°F)
Physical Form: Powder, packaged under argon
Density: 0.865
Sensitivity: Moisture Sensitive
Appearance: White to pale brown
Melting Point: 87-89°
Boiling Point: 90°/0.05mm subl.
UN No.: UN3131
MDL No.: MFCD00269811
Applications: Preparation of aluminum nitrides
Vzorec:
C₁₂H₃₆Al₂N₆ MW: 318,42 g/mol Bod varu: 90 °C (0,05 mmHg) (subl.) Bod tání: 87…89 °C Hustota: 0,865 Bod vzplanutí: 21 °C (70 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00269811 CAS číslo: 32093-39-3 UN: 3131 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...