Tisk…

Lithium niobium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43175.AEEA 11120 CZK
43175.AE 43175.AC
Lithium niobium isopropoxide 5% (w/v) in isopropanol (IPA) , 99% (metals basis)
Lithium niobium isopropoxide

Purity: 99+% CAS No.: 21864-21-1 Molecular Formula: C18H42LiNbO6 Formula Weight: 454.38 Physical Form: Liquid Sensitivity: Moisture Sensitive Odour: Alcohol-like UN No.: UN1219 MDL No.: MFCD00210626

Vzorec: LiNb(OC₃H₇)₆
MW: 454.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210626
CAS číslo: 21864-21-1
UN: 1219
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR