Tisk…
Paladium 5% on calcium carbonate for use as a catalyst lead poisoned Type A306060-5
Dodavatel: Thermo Fisher Scientific
Nebezpečí
43172.14 43172.06 43172.22
43172.14EA 14210 CZK
43172.14 43172.06 43172.22
Paladium 5% on calcium carbonate for use as a catalyst lead poisoned Type A306060-5
Palladium
CAS No.: 7440-05-3 Physical Form: Powder UN No.: UN3077 MDL No.: MFCD03427452
Applications: Selective hydrogenation
Vzorec: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS číslo: 7440-05-3
UN: 3077
ADR: 9 III