Tisk…

Ruthenium(III) nitrosyl chloride, hydrate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Trichloronitrosylruthenium(III), Ruthenium(III) nitrosylchloride

43089.03 43089.06
43089.03EA 5190 CZK
43089.03 43089.06
Ruthenium(III) nitrosyl chloride, hydrate
Ruthenium(III) nitrosyl chloride

CAS No.: 32535-76-5 Molecular Formula: Ru(NO)Cl3.xH2O Formula Weight: 237.44(anhy) Physical Form: Crystalline Melting Point: 180° dec. MDL No.: MFCD00049765

Vzorec: Ru(NO)Cl₃
MW: 237.44 g/mol (anhy)
Bod tání: 180 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00049765
CAS číslo: 32535-76-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR