Tisk…

Poly(vinylbenzyl chloride)

Dodavatel: Thermo Fisher Scientific
Varování

43067.09 43067.18 43067.04
43067.09EA 4400 CZK
43067.09 43067.18 43067.04
Poly(vinylbenzyl chloride)
Poly(vinylbenzyl chloride)

CAS No.: 9080-67-5 Molecular Formula: -[CH2CH(C6H4CH2Cl-]n Physical Form: Solid MDL No.: MFCD00135445

Vzorec: -[CH₂CH(C₆H₄CH₂Cl-]n
Storage Temperature: Ambient
MDL Number: MFCD00135445
CAS číslo: 9080-67-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR