Tisk…
Tetramethylammonium formate 30% (w/w) vodný roztok
Dodavatel: Thermo Scientific
42710.AE 42710.AP
42710.AEEA 2800 CZK
42710.AE 42710.AP
Tetramethylammonium formate 30% (w/w) vodný roztok
Tetramethylammonium formate
CAS No.: 59138-84-0 Molecular Formula: C5H13NO2 Formula Weight: 119.16 Physical Form: Liquid Beilstein Reference: 3916177 MDL No.: MFCD00054402
Vzorec: C₅H₁₃NO₂
MW: 119,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054402
CAS číslo: 59138-84-0
EINECS: 261-622-8