Tisk…
Palladium hydroxide on carbon (Pearlman's catalyst) on carbon (Pearlman's catalyst) wetted with ca. 50% water
Dodavatel: Thermo Fisher Scientific
42578.04 42578.09 42578.18
42578.04EA 5530 CZK
42578.04 42578.09 42578.18
Palladium hydroxide on carbon (Pearlman's catalyst) on carbon (Pearlman's catalyst) wetted with ca. 50% water
Palladium hydroxide
CAS No.: 12135-22-7 Odour: Odourless MDL No.: MFCD00064599
Applications: Hydrogenolysis of benzyl-nitrogen bonds
Vzorec: H₂O₂Pd
MW: 140.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS číslo: 12135-22-7
EINECS: 235-219-2