Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
42527.03
42527.06
42527.03EA
6970
CZK
42527.03
42527.06
Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)
Dihydrogen hexabromoosmate(IV) hydrate
Purity: 99.9%
Molecular Formula: H2OsBr6.xH2O
Formula Weight: 352.02(anhy)
Physical Form: Crystalline
Appearance: Grey-black
Melting Point: dec.
UN No.: UN3260
MDL No.: MFCD01863474
Vzorec:
H₂OsBr₆·xH₂O MW: 352.02 g/mol (anhy) Bod tání: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD01863474 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...