Lanthanum(III) 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
42522.AC
42522.AE
42522.ACEA
6800
CZK
42522.AC
42522.AE
Lanthanum(III) 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99.9% (REO, rare earth oxide basis)
Lanthanum(III) 2-methoxyethoxide
Purity: 99.9%
CAS No.: 115685-56-8
Molecular Formula: C9H21LaO6
Formula Weight: 364.17
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Yellow
Boiling Point: 124°
UN No.: UN1188
MDL No.: MFCD00210619
Applications: Preparation of lanthanum oxide thin films by the Sol-Gel method
Vzorec:
C₉H₂₁LaO₆ MW: 364.17 g/mol Bod varu: 124 °C Hustota: 0.96 Bod vzplanutí: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210619 CAS číslo: 115685-56-8 UN: 1188 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...