3-Mercaptopropyl(dimethoxy)methylsilane ≥95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
42426.AD
42426.AK
42426.ADEA
1880
CZK
42426.AD
42426.AK
3-Mercaptopropyl(dimethoxy)methylsilane ≥95%
3-Mercaptopropyl(dimethoxy)methylsilane
Purity: 95%
CAS No.: 31001-77-1
Molecular Formula: C6H16O2SSi
Formula Weight: 180.34
Flash Point: 93°(199°F)
Physical Form: Liquid
Density: 1.000
Refractive Index: 1.4500
Sensitivity: Moisture Sensitive
Appearance: Colourless
Boiling Point: 96°/30mm
MDL No.: MFCD00053923
Vzorec:
C₆H₁₆O₂SSi Bod varu: 96 °C (30 mmHg) Hustota: 1,000 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053923 CAS číslo: 31001-77-1 EINECS: 250-426-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...