Indium(III) acetate ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
42230.09
42230.18
42230.09EA
3080
CZK
42230.09
42230.18
Indium(III) acetate ≥99.99% (metals basis)
Indium(III) acetate
Purity: 99.99%
CAS No.: 25114-58-3
Molecular Formula: In(OOCCH3)3
Formula Weight: 291.96
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00054347
Vzorec:
C₂H₄InO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00054347 CAS číslo: 25114-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...