Tisk…

Platina 40% on carbon, HiSPEC® 4000

Dodavatel: Thermo Fisher Scientific
Nebezpečí

42204.03 42204.14
42204.03EA 0 CZK
42204.03 42204.14
Platina 40% on carbon, HiSPEC® 4000
Platina

CAS No.: 7440-06-4 Physical Form: Powder UN No.: UN1325 MDL No.: MFCD00011179

Vzorec: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
UN: 1325
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR